Assay Details
| Gene Symbol: IL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003301.1 | NP_001003301.1 | 4 - 5 | 514 | 66 | |
| GenBank mRNA | AF275796.1 | - | 4 - 5 | 457 | 66 | |
| U12234.1 | - | 4 - 5 | 514 | 66 | ||
| Z70045.1 | - | 3 - 4 | 438 | 66 | ||
More Information
Set Membership: |
|

IL6
Genomic Map