Assay Details
| Gene Symbol: IL13RA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003075.1 | NP_001003075.1 | 5 - 6 | 702 | 72 | |
| GenBank mRNA | AF314533.1 | - | 5 - 6 | 702 | 72 | |
| DQ195087.1 | - | 4 - 5 | 521 | 72 | ||
More Information
Set Membership: |
|

IL13RA2
Genomic Map