Assay Details
| Gene Symbol: IL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003077.1 | NP_001003077.1 | 386 | 89 | ||
| GenBank mRNA | AF328930.1 | - | 250 | 83 | ||
| AF333120.1 | - | 146 | 83 | |||
| EU426968.1 | - | 379 | 83 | |||
| U33843.1 | - | 386 | 89 | |||
More Information
Set Membership: |
|

IL10
Genomic Map