Assay Details
| Gene Symbol: CLDN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003088.1 | NP_001003088.1 | 946 | 88 | ||
| GenBank mRNA | AF358908.1 | - | 946 | 88 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CLDN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003088.1 | NP_001003088.1 | 946 | 88 | ||
| GenBank mRNA | AF358908.1 | - | 946 | 88 | ||
Set Membership: |
|