Assay Details
| Gene Symbol: RET | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197099.1 | NP_001184028.1 | 5 - 6 | 1041 | 65 | |
| XM_014108752.2 | XP_013964227.1 | 5 - 6 | 1110 | 65 | ||
| XM_022411580.1 | XP_022267288.1 | 5 - 6 | 1085 | 65 | ||
| GenBank mRNA | AF364316.1 | - | 1 - 2 | 157 | 65 | |
More Information
Set Membership: |
|

RET
Genomic Map