Assay Details
| Gene Symbol: MYL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003069.1 | NP_001003069.1 | 302 | 119 | ||
| GenBank mRNA | AF440218.1 | - | 302 | 119 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MYL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003069.1 | NP_001003069.1 | 302 | 119 | ||
| GenBank mRNA | AF440218.1 | - | 302 | 119 | ||
Set Membership: |
|