Assay Details
| Gene Symbol: ELANE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003378.2 | NP_001003378.1 | 4 - 5 | 626 | 98 | |
| GenBank mRNA | AF494190.1 | - | 4 - 5 | 629 | 98 | |
More Information
Set Membership: |
|

ELANE
Genomic Map