Assay Details
| Gene Symbol: TBXT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003092.1 | NP_001003092.1 | 1 - 2 | 201 | 75 | |
| GenBank mRNA | AJ245513.1 | - | 1 - 2 | 201 | 75 | |
More Information
Set Membership: |
|

TBXT
Genomic Map