Assay Details
| Gene Symbol: RHPN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003008.1 | NP_001003008.1 | 7 - 8 | 779 | 56 | |
| GenBank mRNA | AJ347749.1 | - | 7 - 8 | 779 | 56 | |
More Information
Set Membership: |
|

RHPN2
Genomic Map