Assay Details
| Gene Symbol: RPS14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287105.1 | NP_001274034.1 | 4 - 5 | 481 | 73 | |
| NM_001287106.1 | NP_001274035.1 | 4 - 5 | 452 | 73 | ||
| GenBank mRNA | AJ388514.1 | - | 2 - 3 | 168 | 73 | |
| CO658560.1 | - | 1 - 2 | 129 | 73 | ||
| DN863506.1 | - | 1 - 2 | 144 | 73 | ||
| DN871196.1 | - | 4 - 5 | 452 | 73 | ||
| DN872809.1 | - | 2 | 317 | 73 | ||
| GR889127.1 | - | 4 - 5 | 482 | 73 | ||
| GR894797.1 | - | 4 - 5 | 430 | 73 | ||
More Information
Set Membership: |
|

RPS14
Genomic Map