Assay Details
| Gene Symbol: RPS18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048082.1 | NP_001041547.1 | 4 - 5 | 332 | 101 | |
| GenBank mRNA | AJ388519.1 | - | 2 - 3 | 123 | 101 | |
More Information
Set Membership: |
|

RPS18
Genomic Map