Assay Details
| Gene Symbol: GJB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197148.1 | NP_001184077.1 | 529 | 139 | ||
| GenBank mRNA | AJ439693.1 | - | 194 | 139 | ||
| DN870952.1 | - | 213 | 139 | |||
| DN905693.1 | - | 529 | 139 | |||
More Information
Set Membership: |
|

GJB2
Genomic Map