Assay Details
| Gene Symbol: ASB17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003018.2 | NP_001003018.2 | 1 - 2 | 507 | 111 | |
| GenBank mRNA | AJ518867.1 | - | 1 - 2 | 514 | 111 | |
More Information
Set Membership: |
|

ASB17
Genomic Map