Assay Details
Gene Symbol: DEFB122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024641.2 | NP_001019812.2 | 1 - 2 | 189 | 64 | |
NM_001313788.1 | NP_001300717.1 | 1 - 2 | 189 | 64 | ||
NR_132650.1 | 1 - 2 | 189 | 64 | |||
GenBank mRNA | AY169790.1 | - | 1 - 2 | 111 | 64 | |
AY169792.1 | - | 1 - 2 | 111 | 64 | ||
DQ011991.1 | - | 1 - 2 | 189 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
