Assay Details
| Gene Symbol: HRH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005635258.2 | XP_005635315.2 | 2 | 1006 | 85 | |
| GenBank mRNA | AH012621.2 | - | 3 | 828 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: HRH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005635258.2 | XP_005635315.2 | 2 | 1006 | 85 | |
| GenBank mRNA | AH012621.2 | - | 3 | 828 | 85 | |
Set Membership: |
|