Assay Details
| Gene Symbol: KCNE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194963.1 | NP_001181892.1 | 1 | 226 | 99 | |
| GenBank mRNA | AY254370.1 | - | 1 | 74 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: KCNE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194963.1 | NP_001181892.1 | 1 | 226 | 99 | |
| GenBank mRNA | AY254370.1 | - | 1 | 74 | 99 | |
Set Membership: |
|