Assay Details
| Gene Symbol: OAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048131.1 | NP_001041596.1 | 945 | 61 | ||
| XM_005636024.3 | XP_005636081.1 | 999 | 61 | |||
| GenBank mRNA | AY863104.1 | - | 945 | 61 | ||
More Information
Set Membership: |
|

OAS1
Genomic Map