Assay Details
| Gene Symbol: ADRA2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022411624.1 | XP_022267332.1 | 1 | 1363 | 88 | |
| GenBank mRNA | DQ009035.1 | - | 1 | 119 | 88 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ADRA2A
Genomic Map