Assay Details
| Gene Symbol: SST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003307.1 | NP_001003307.1 | 1 - 2 | 216 | 84 | |
| GenBank mRNA | L42325.1 | - | 1 - 2 | 216 | 84 | |
More Information
Set Membership: |
|

SST
Genomic Map