Assay Details
| Gene Symbol: GNGT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003225.1 | NP_001003225.1 | 1 - 2 | 60 | 114 | |
| GenBank mRNA | Z75156.1 | - | 1 - 2 | 60 | 114 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001003225.1, Z75156.1 transcripts.
Set Membership: |
|

GNGT1
Genomic Map