Assay Details
| Gene Symbol: CYP19A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008715.1 | NP_001008715.1 | 2 - 3 | 300 | 80 | |
| XM_022412491.1 | XP_022268199.1 | 3 - 4 | 357 | 80 | ||
| XM_022412492.1 | XP_022268200.1 | 4 - 5 | 795 | 80 | ||
| XM_022412493.1 | XP_022268201.1 | 4 - 5 | 736 | 80 | ||
| XM_022412494.1 | XP_022268202.1 | 3 - 4 | 553 | 80 | ||
| XM_022412495.1 | XP_022268203.1 | 3 - 4 | 356 | 80 | ||
| XM_022412496.1 | XP_022268204.1 | 2 - 3 | 239 | 80 | ||
| XM_022412497.1 | XP_022268205.1 | 2 - 3 | 250 | 80 | ||
| XM_022412498.1 | XP_022268206.1 | 2 - 3 | 235 | 80 | ||
| GenBank mRNA | AJ854107.1 | - | 2 - 3 | 300 | 80 | |
More Information
Set Membership: |
|

CYP19A1
Genomic Map