Assay Details
| Gene Symbol: B3GAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013419.1 | NP_001013437.1 | 1 - 2 | 609 | 73 | |
| GenBank mRNA | AJ888982.1 | - | 1 - 2 | 609 | 73 | |
More Information
Set Membership: |
|

B3GAT2
Genomic Map