Assay Details
| Gene Symbol: COMMD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003055.1 | NP_001003055.1 | 2 - 3 | 476 | 87 | |
| GenBank mRNA | AX786408.1 | - | 2 - 3 | 476 | 87 | |
| AY047597.1 | - | 2 - 3 | 476 | 87 | ||
More Information
Set Membership: |
|

COMMD1
Genomic Map