Assay Details
| Gene Symbol: KITLG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012735.2 | NP_001012753.1 | 4 - 5 | 549 | 157 | |
| XM_022427652.1 | XP_022283360.1 | 4 - 5 | 560 | 157 | ||
| GenBank mRNA | AY094361.1 | - | 3 - 4 | 308 | 157 | |
| S53329.1 | - | 3 - 4 | 355 | 157 | ||
More Information
Set Membership: |
|

KITLG
Genomic Map