Assay Details
| Gene Symbol: MCHR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003037.1 | NP_001003037.1 | 3 - 4 | 592 | 105 | |
| GenBank mRNA | AY112659.1 | - | 3 - 4 | 592 | 105 | |
More Information
Set Membership: |
|

MCHR2
Genomic Map