Assay Details
| Gene Symbol: HMGB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002937.2 | NP_001002937.1 | 316 | 102 | ||
| XM_022409535.1 | XP_022265243.1 | 323 | 102 | |||
| GenBank mRNA | AY135519.1 | - | 238 | 102 | ||
| DN349572.1 | - | 2 | 316 | 102 | ||
More Information
Set Membership: |
|

HMGB1
Genomic Map