Assay Details
| Gene Symbol: KLRA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002986.1 | NP_001002986.1 | 6 - 7 | 815 | 153 | |
| GenBank mRNA | AY191818.1 | - | 6 - 7 | 815 | 153 | |
More Information
Set Membership: |
|

KLRA1
Genomic Map