Assay Details
| Gene Symbol: SLC46A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002997.1 | NP_001002997.1 | 1 - 2 | 1220 | 76 | |
| GenBank mRNA | AY195876.1 | - | 1 - 2 | 1220 | 76 | |
More Information
Set Membership: |
|

SLC46A2
Genomic Map