Assay Details
| Gene Symbol: SLC46A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002997.1 | NP_001002997.1 | 2 - 3 | 1308 | 71 | |
| NR_120617.1 | 2 - 3 | 1136 | 71 | |||
| GenBank mRNA | AY195876.1 | - | 2 - 3 | 1308 | 71 | |
| AY225149.1 | - | 2 - 3 | 1136 | 71 | ||
More Information
Set Membership: |
|

SLC46A2
Genomic Map