Assay Details
| Gene Symbol: KRT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003392.1 | NP_001003392.1 | 8 - 9 | 1512 | 63 | |
| GenBank mRNA | AY318945.1 | - | 8 - 9 | 1512 | 63 | |
More Information
Set Membership: |
|

KRT1
Genomic Map