Assay Details
| Gene Symbol: AKAP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007123.1 | NP_001007124.1 | 10 - 11 | 1305 | 68 | |
| GenBank mRNA | AY382602.1 | - | 10 - 11 | 1305 | 68 | |
More Information
Set Membership: |
|

AKAP8
Genomic Map