Assay Details
| Gene Symbol: HAMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007140.1 | NP_001007141.1 | 1 - 2 | 114 | 57 | |
| GenBank mRNA | AY590589.1 | - | 1 - 2 | 92 | 57 | |
| AY772532.1 | - | 1 - 2 | 114 | 57 | ||
| AY899807.1 | - | 1 - 2 | 92 | 57 | ||
More Information
Set Membership: |
|

HAMP
Genomic Map