Assay Details
| Gene Symbol: REN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003194.2 | NP_001003194.1 | 8 - 9 | 1043 | 65 | |
| XM_005640752.2 | XP_005640809.1 | 9 - 10 | 1130 | 65 | ||
| XM_022415287.1 | XP_022270995.1 | 9 - 10 | 1148 | 65 | ||
| GenBank mRNA | AY630442.1 | - | 8 - 9 | 1043 | 65 | |
More Information
Set Membership: |
|

REN
Genomic Map