Assay Details
| Gene Symbol: P2RX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639268.4 | XP_003639316.2 | 8 - 9 | 1044 | 88 | |
| XM_014116853.2 | XP_013972328.1 | 8 - 9 | 1045 | 88 | ||
| GenBank mRNA | AY708380.1 | - | 2 - 3 | 208 | 88 | |
More Information
Set Membership: |
|

P2RX5
Genomic Map