Assay Details
| Gene Symbol: CBLIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005759.1 | NP_001005759.1 | 4 - 5 | 549 | 87 | |
| GenBank mRNA | AY730674.1 | - | 4 - 5 | 549 | 87 | |
More Information
Set Membership: |
|

CBLIF
Genomic Map