Assay Details
| Gene Symbol: VHL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008552.1 | NP_001008552.1 | 505 | 74 | ||
| XM_014121534.2 | XP_013977009.1 | 654 | 74 | |||
| GenBank mRNA | AY764285.1 | - | 505 | 74 | ||
| GU563722.1 | - | 252 | 74 | |||
More Information
Set Membership: |
|

VHL
Genomic Map