Assay Details
| Gene Symbol: CFTR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007143.1 | NP_001007144.1 | 8 - 9 | 1120 | 129 | |
| GenBank mRNA | AY780429.1 | - | 8 - 9 | 1120 | 129 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CFTR
Genomic Map