Assay Details
| Gene Symbol: RHCE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048036.2 | NP_001041501.1 | 1 - 2 | 214 | 67 | |
| GenBank mRNA | AY831676.1 | - | 1 - 2 | 214 | 67 | |
More Information
Set Membership: |
|

RHCE
Genomic Map