Assay Details
| Gene Symbol: RHCE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048036.2 | NP_001041501.1 | 4 - 5 | 696 | 98 | |
| GenBank mRNA | AY831676.1 | - | 4 - 5 | 696 | 98 | |
More Information
Set Membership: |
|

RHCE
Genomic Map