Assay Details
| Gene Symbol: MACO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038648.1 | NP_001033737.1 | 8 - 9 | 1603 | 63 | |
| GenBank mRNA | AY845020.1 | - | 8 - 9 | 1603 | 63 | |
More Information
Set Membership: |
|

MACO1
Genomic Map