Assay Details
| Gene Symbol: UOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011886.1 | NP_001011886.1 | 3 - 4 | 392 | 79 | |
| GenBank mRNA | AY871313.1 | - | 3 - 4 | 392 | 79 | |
More Information
Set Membership: |
|

UOX
Genomic Map