Assay Details
| Gene Symbol: TAS1R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031819.1 | NP_001026989.1 | 2 - 3 | 479 | 59 | |
| GenBank mRNA | AY916758.1 | - | 2 - 3 | 479 | 59 | |
More Information
Set Membership: |
|

TAS1R2
Genomic Map