Assay Details
| Gene Symbol: TAS1R3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031821.1 | NP_001026991.1 | 483 | 81 | ||
| GenBank mRNA | AY916759.1 | - | 483 | 81 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: TAS1R3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031821.1 | NP_001026991.1 | 483 | 81 | ||
| GenBank mRNA | AY916759.1 | - | 483 | 81 | ||
Set Membership: |
|