Assay Details
| Gene Symbol: PODXL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020809.1 | NP_001018645.1 | 6 - 7 | 1358 | 73 | |
| GenBank mRNA | AY970669.1 | - | 6 - 7 | 1358 | 73 | |
More Information
Set Membership: |
|

PODXL
Genomic Map