Assay Details
| Gene Symbol: CPB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003005.2 | NP_001003005.2 | 10 - 11 | 1079 | 69 | |
| GenBank mRNA | D78348.1 | - | 10 - 11 | 1066 | 69 | |
| DN360182.1 | - | 5 - 6 | 557 | 69 | ||
More Information
Set Membership: |
|

CPB1
Genomic Map