Assay Details
| Gene Symbol: GUCY1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018035.1 | NP_001018045.1 | 1 - 2 | 37 | 69 | |
| GenBank mRNA | DQ008575.1 | - | 1 - 2 | 37 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001018035.1 transcripts.
Set Membership: |
|

GUCY1A1
Genomic Map