Assay Details
| Gene Symbol: GUCY1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018035.1 | NP_001018045.1 | 6 - 7 | 1443 | 92 | |
| XM_005628839.3 | XP_005628896.1 | 6 - 7 | 1731 | 92 | ||
| XM_005628841.3 | XP_005628898.1 | 5 - 6 | 1501 | 92 | ||
| XM_005628842.3 | XP_005628899.1 | 5 - 6 | 1264 | 92 | ||
| XM_014119231.2 | XP_013974706.1 | 6 - 7 | 1652 | 92 | ||
| XM_022427665.1 | XP_022283373.1 | 6 - 7 | 1442 | 92 | ||
| XM_022427666.1 | XP_022283374.1 | 6 - 7 | 1439 | 92 | ||
| XM_022427667.1 | XP_022283375.1 | 5 - 6 | 1438 | 92 | ||
| XM_022427668.1 | XP_022283376.1 | 5 - 6 | 1277 | 92 | ||
| GenBank mRNA | DQ008575.1 | - | 6 - 7 | 1443 | 92 | |
More Information
Set Membership: |
|

GUCY1A1
Genomic Map