Assay Details
| Gene Symbol: GUCY1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018034.2 | NP_001018044.1 | 1 - 2 | 102 | 132 | |
| GenBank mRNA | DQ008576.1 | - | 1 - 2 | 102 | 132 | |
More Information
Set Membership: |
|

GUCY1B1
Genomic Map