Assay Details
| Gene Symbol: IL18BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048018.1 | NP_001041483.1 | 2 - 3 | 226 | 58 | |
| XM_005633286.3 | XP_005633343.1 | 3 - 4 | 1396 | 58 | ||
| XM_014122423.2 | XP_013977898.1 | 3 - 4 | 1396 | 58 | ||
| XM_022407424.1 | XP_022263132.1 | 2 - 3 | 374 | 58 | ||
| GenBank mRNA | DQ195088.1 | - | 2 - 3 | 226 | 58 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001048018.1 transcripts.
Set Membership: |
|

IL18BP
Genomic Map