Assay Details
| Gene Symbol: PODXL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014121652.2 | XP_013977127.1 | 1456 | 69 | ||
| GenBank mRNA | DQ202369.1 | - | 1280 | 69 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PODXL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014121652.2 | XP_013977127.1 | 1456 | 69 | ||
| GenBank mRNA | DQ202369.1 | - | 1280 | 69 | ||
Set Membership: |
|